Learn More
3,4,5-Trimethoxybenzaldehyde, 98%
CAS: 86-81-7 | C10H12O4 | 196.202 g/mol
Supplier: Thermo Scientific Chemicals A1503822
Description
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.
Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
| 3,4,5-Trimethoxybenzaldehyde | |
| 73°C to 77°C | |
| C10H12O4 | |
| 100 g | |
| Air Sensitive | |
| Soluble in water (1.49mg/L at 25°C),alcohol,toluene,acetone,and methanol. | |
| COC1=CC(=CC(=C1OC)OC)C=O | |
| 196.202 | |
| 196.2 |
| 86-81-7 | |
| 163°C to 165°C (10 mmHg) | |
| MFCD00003364 | |
| 395163 | |
| benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde | |
| OPHQOIGEOHXOGX-UHFFFAOYSA-N | |
| 3,4,5-trimethoxybenzaldehyde | |
| 6858 | |
| 98% |
Chemical Identifiers
| 86-81-7 | |
| 196.202 | |
| OPHQOIGEOHXOGX-UHFFFAOYSA-N | |
| 3,4,5-trimethoxybenzaldehyde |
| C10H12O4 | |
| MFCD00003364 | |
| 6858 | |
| COC1=CC(=CC(=C1OC)OC)C=O |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 201-701-6
RTECSNumber : CU8462000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only