missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde, 98%

Catalog No. AAA1503836
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1503836 500 g
AAA1503814 25 g
AAA1503822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1503836 Supplier Thermo Scientific Chemicals Supplier No. A1503836

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 86-81-7 | C10H12O4 | 196.202 g/mol

3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 86-81-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003364
InChI Key OPHQOIGEOHXOGX-UHFFFAOYSA-N
PubChem CID 6858
IUPAC Name 3,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)OC)C=O

Specifications

Melting Point 73°C to 77°C
Boiling Point 163°C to 165°C (10 mmHg)
Quantity 500 g
Beilstein 395163
Sensitivity Air Sensitive
Synonym benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde
Solubility Information Soluble in water (1.49mg/L at 25°C),alcohol,toluene,acetone,and methanol.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 201-701-6
RTECSNumber CU8462000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only