missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7136058
Change view
Click to view available options
Quantity:
25 g
100 g
Catalog No. Quantity
T005925G 25 g
T0059100G 100 g
2 options

Catalog No. T005925G

Supplier: TCI America T005925G

Only null left

Chemical Identifiers

CAS 86-81-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003364
InChI Key OPHQOIGEOHXOGX-UHFFFAOYSA-N
PubChem CID 6858
IUPAC Name 3,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)OC)C=O

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 165°C
Quantity 25 g
Synonym benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde
Formula Weight 196.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxybenzaldehyde
EINECSNumber (3)-2920
RTECSNumber CU8462000
TSCA Yes