missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyaniline 98.0+%, TCI America™

Catalog No. T10555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10555G 5 g
T105525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10555G Supplier TCI America Supplier No. T10555G

Chemical Identifiers

CAS 24313-88-0
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00008393
InChI Key XEFRNCLPPFDWAC-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
PubChem CID 32285
IUPAC Name 3,4,5-trimethoxyaniline
SMILES COC1=CC(=CC(=C1OC)OC)N

Specifications

Melting Point 112°C
Quantity 5 g
Formula Weight 183.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trimethoxyaniline