missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyaniline, 98+%

Catalog No. AAL0161914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0161914 25 g
AAL0161906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0161914 Supplier Thermo Scientific Chemicals Supplier No. L0161914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24313-88-0 | C9H13NO3 | 183.207 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24313-88-0
Molecular Formula C9H13NO3
Molecular Weight (g/mol) 183.207
MDL Number MFCD00008393
InChI Key XEFRNCLPPFDWAC-UHFFFAOYSA-N
Synonym benzenamine, 3,4,5-trimethoxy, 3,4,5-trimethoxy aniline, 3,4,5-trimethoxybenzenamine, 3,4,5-trimethoxy-phenylamine, 3,4,5-trimethoxyphenylamine, 3,5-trimethoxyaniline, 3,4,5-trimethoxyanilin, acmc-1cdy7, intermediates-zcf02654, 3,4,5-trimethoxyaniline
PubChem CID 32285
IUPAC Name 3,4,5-trimethoxyaniline
SMILES COC1=CC(=CC(=C1OC)OC)N

Specifications

Melting Point 110°C to 113°C
Quantity 25 g
UN Number UN2811
Beilstein 642919
Formula Weight 183.21
Percent Purity ≥98%
Chemical Name or Material 3,4,5-Trimethoxyaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 246-154-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only