missing translation for 'onlineSavingsMsg'
Learn More

3',4',5'-Trimethoxyacetophenone 98.0+%, TCI America™

Catalog No. p-7136483
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08985G 5 g
T089825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08985G Supplier TCI America Supplier No. T08985G

Chemical Identifiers

CAS 1136-86-3
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00008738
InChI Key VUGQIIQFXCXZJU-UHFFFAOYSA-N
Synonym 3',4',5'-trimethoxyacetophenone, 1-3,4,5-trimethoxyphenyl ethanone, 3,4,5-trimethoxyacetophenone, ethanone, 1-3,4,5-trimethoxyphenyl, 1-3,4,5-trimethoxyphenyl ethan-1-one, 3,4,5-trimethoxy acetophenone, pubchem22820, acmc-1c5os, 3,5-trimethoxy acetophenone, 1-acetyl-3,4,5-trimethoxybenzene
PubChem CID 14345
ChEBI CHEBI:86547
IUPAC Name 1-(3,4,5-trimethoxyphenyl)ethan-1-one
SMILES COC1=CC(=CC(OC)=C1OC)C(C)=O

Specifications

Melting Point 82°C
Boiling Point 224°C
Quantity 5 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′,4′,5′-Trimethoxyacetophenone