missing translation for 'onlineSavingsMsg'
Learn More

3',4',5'-Trimethoxyacetophenone, 99%

Catalog No. p-7058313
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0643906 5 g
AAL0643914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0643906 Supplier Thermo Scientific Chemicals Supplier No. L0643906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1136-86-3 | C11H14O4 | 210.23 g/mol

3',4',5'-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′,5′-Trimethoxyacetophenone is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Slightly soluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases. Store away from strong oxidizing agents and stong bases. Avoid excessive heat and light exposure.

Chemical Identifiers

CAS 1136-86-3
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00008738
InChI Key VUGQIIQFXCXZJU-UHFFFAOYSA-N
Synonym 3',4',5'-trimethoxyacetophenone, 1-3,4,5-trimethoxyphenyl ethanone, 3,4,5-trimethoxyacetophenone, ethanone, 1-3,4,5-trimethoxyphenyl, 1-3,4,5-trimethoxyphenyl ethan-1-one, 3,4,5-trimethoxy acetophenone, pubchem22820, acmc-1c5os, 3,5-trimethoxy acetophenone, 1-acetyl-3,4,5-trimethoxybenzene
PubChem CID 14345
ChEBI CHEBI:86547
IUPAC Name 1-(3,4,5-trimethoxyphenyl)ethanone
SMILES COC1=CC(=CC(OC)=C1OC)C(C)=O

Specifications

Melting Point 79°C to 83°C
Boiling Point 173°C to 174°C (10 mmHg)
Quantity 5 g
Beilstein 1463133
Solubility Information Slightly soluble in water.
Formula Weight 210.23
Percent Purity 99%
Chemical Name or Material 3',4',5'-Trimethoxyacetophenone
EINECSNumber 214-501-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only