missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4',5-Trimethoxy-trans-stilbene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 57°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 270.33 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3,4′,5-Trimethoxy-trans-stilbene |
Safety and Handling
| TSCA | No |