missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzaldehyde 98.0+%, TCI America™

Catalog No. p-7137337
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26501G 1 g
T26505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26501G Supplier TCI America Supplier No. T26501G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13677-79-7
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003371
InChI Key RGZHEOWNTDJLAQ-UHFFFAOYSA-N
Synonym gallaldehyde, benzaldehyde, 3,4,5-trihydroxy, pyrogallol-5-carboxaldehyde, 3,4,5-trihydroxy benzaldehyde, pubchem21300, 3,4,5-trihydroxybenz, acmc-209c9s, 3,4,5-trihydroxybenzaldhyde, 3,4,5-trihydroxy-benzaldehyde, 3,4,5-tris oxidanyl benzaldehyde
PubChem CID 83651
IUPAC Name 3,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1O

Specifications

Melting Point 215°C
Quantity 1 g
Formula Weight 154.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Trihydroxybenzaldehyde