missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxybenzaldehyde hydrate, 97%

Catalog No. AAA1927003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1927003 1 g
AAA1927006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1927003 Supplier Thermo Scientific Chemicals Supplier No. A1927003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13677-79-7 | C7H6O4 | 154.12 g/mol

3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-trihydroxybenzaldehyde from Geum japonicum has dual inhibitory effect on matrix metalloproteinase 9; inhibition of gelatinoytic activity as well as MMP-9 expression in TNF-α induced HASMC.

Solubility
Soluble in DMSO and Methanol.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 13677-79-7
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00003371
InChI Key RGZHEOWNTDJLAQ-UHFFFAOYSA-N
Synonym gallaldehyde, benzaldehyde, 3,4,5-trihydroxy, pyrogallol-5-carboxaldehyde, 3,4,5-trihydroxy benzaldehyde, pubchem21300, 3,4,5-trihydroxybenz, acmc-209c9s, 3,4,5-trihydroxybenzaldhyde, 3,4,5-trihydroxy-benzaldehyde, 3,4,5-tris oxidanyl benzaldehyde
PubChem CID 83651
IUPAC Name 3,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1O

Specifications

Melting Point ∼212°C (decomposition)
Quantity 1 g
Beilstein 2091654
Sensitivity Air Sensitive
Solubility Information Soluble in DMSO and Methanol.
Formula Weight 154.12 (Anhydrous)
Percent Purity 97%
Chemical Name or Material 3,4,5-Trihydroxybenzaldehyde hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-168-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only