missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trichloropyridine 98.0+%, TCI America™

Supplier:  TCI America T271425G

 View more versions of this product

Catalog No. T271425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3,4,5-Trichloropyridine
73°C
215°C
MFCD00051685
pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g
C1=C(C(=C(C=N1)Cl)Cl)Cl
182.428
182.43
Crystalline Powder
33216-52-3
White-Yellow
C5H2Cl3N
25 g
KKWRVUBDCJQHBZ-UHFFFAOYSA-N
3,4,5-trichloropyridine
3786531
≥98.0% (GC,T)

Chemical Identifiers

33216-52-3
182.428
KKWRVUBDCJQHBZ-UHFFFAOYSA-N
3786531
C1=C(C(=C(C=N1)Cl)Cl)Cl
C5H2Cl3N
MFCD00051685
pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g
3,4,5-trichloropyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions