missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4,5-Trichloropyridine 98.0+%, TCI America™
$55.84 - $221.39
Chemical Identifiers
| CAS | 33216-52-3 |
|---|---|
| Molecular Formula | C5H2Cl3N |
| Molecular Weight (g/mol) | 182.428 |
| MDL Number | MFCD00051685 |
| InChI Key | KKWRVUBDCJQHBZ-UHFFFAOYSA-N |
| Synonym | pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g |
| PubChem CID | 3786531 |
| IUPAC Name | 3,4,5-trichloropyridine |
| SMILES | C1=C(C(=C(C=N1)Cl)Cl)Cl |
Chemical Identifiers
| 33216-52-3 | |
| 182.428 | |
| KKWRVUBDCJQHBZ-UHFFFAOYSA-N | |
| 3786531 | |
| C1=C(C(=C(C=N1)Cl)Cl)Cl |
| C5H2Cl3N | |
| MFCD00051685 | |
| pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g | |
| 3,4,5-trichloropyridine |
Specifications
| 33216-52-3 | |
| White-Yellow | |
| C5H2Cl3N | |
| 5 g | |
| KKWRVUBDCJQHBZ-UHFFFAOYSA-N | |
| 3,4,5-trichloropyridine | |
| 3786531 | |
| ≥98.0% (GC,T) | |
| 3,4,5-Trichloropyridine |
| 73°C | |
| 215°C | |
| MFCD00051685 | |
| pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g | |
| C1=C(C(=C(C=N1)Cl)Cl)Cl | |
| 182.428 | |
| 182.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No