missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trichlorobenzotrifluoride 98.0+%, TCI America™

Catalog No. T22815G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22815G 5 g
T228125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22815G Supplier TCI America Supplier No. T22815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50594-82-6
Molecular Formula C7H2Cl3F3
Molecular Weight (g/mol) 249.438
InChI Key FBKFIAIRSQOXJR-UHFFFAOYSA-N
Synonym 1,2,3-trichloro-5-trifluoromethyl benzene, 3,4,5-trichlorobenzotrifluoride, 3,4,5-trichlorotrifluoromethylbenzene, 3,4,5-trichloro-alpha,alpha,alpha-trifluorotoluene, 3,4,5-trichloro-a,a,a-trifluorotoluene, benzene, 1,2,3-trichloro-5-trifluoromethyl, pubchem19737, ksc914i9j, 3,4,5-trichloro benzotrifluoride
PubChem CID 2733399
IUPAC Name 1,2,3-trichloro-5-(trifluoromethyl)benzene
SMILES C1=C(C=C(C(=C1Cl)Cl)Cl)C(F)(F)F

Specifications

Melting Point -6°C
Color Colorless
Boiling Point 113°C
Quantity 5 g
Formula Weight 249.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,4,5-Trichlorobenzotrifluoride