missing translation for 'onlineSavingsMsg'
Learn More

Methyl linoleate, 99%

Catalog No. AC428130050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428130050 5 g Glass bottle
AC428130250 25 g Glass bottle
AC428131000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC428130050 Supplier Thermo Scientific Chemicals Supplier No. 428130050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112-63-0 | C19H34O2 | 294.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-63-0
Molecular Formula C19H34O2
Molecular Weight (g/mol) 294.48
MDL Number MFCD00009534
InChI Key WTTJVINHCBCLGX-NQLNTKRDSA-N
Synonym methyl linoleate, linoleic acid methyl ester, methyl lineoleate, methyl 9z,12z-octadeca-9,12-dienoate, methyl octadecadienoate, methyl 9-cis,12-cis-octadecadienoate, linoleic acid,methyl ester, linoleic acid, methyl ester, methyl linoleate, native, 9,12-octadecadienoic acid z,z-, methyl ester
PubChem CID 5284421
ChEBI CHEBI:69080
IUPAC Name methyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OC

Specifications

Melting Point -35.0°C
Density 0.8900g/mL
Boiling Point 212.0°C (16.0 mmHg)
Flash Point >200°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH=CHCH2CH=CH(CH2)7CO2CH3
Quantity 5 g
Specific Gravity 0.89
Merck Index 15, 6167
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with dmf,fat solvents,oils
Formula Weight 294.48
Percent Purity 99%
Chemical Name or Material Methyl linoleate
Show More Show Less
EINECSNumber 203-993-

RUO – Research Use Only