missing translation for 'onlineSavingsMsg'
Learn More

3,4,5,6-Tetrahydrophthalic anhydride, 98%

Catalog No. p-4509939
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138220250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC138220250 Supplier Thermo Scientific Chemicals Supplier No. 138220250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2426-02-0 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandione
PubChem CID 12475
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 70.0°C to 74.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 17,461
Solubility Information Solubility in water: reacts.
IUPAC Name 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
Formula Weight 152.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5,6-Tetrahydrophthalic anhydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
EINECSNumber 219-374-3

RUO – Research Use Only