accessibility menu, dialog, popup

UserName

CAS RN 2426-02-0

IUPAC Name: 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione
Synonyms: 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: HMMBJOWWRLZEMI-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=C1CCCC2
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)