missing translation for 'onlineSavingsMsg'
Learn More

3,4,4-Trichlorocarbanilide (Triclocarban) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019398
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019398 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019398 Supplier Merck Emd Millipore Supplier No. PHR1303500MG
3,4,4′-Trichlorocarbanilide (Triclocarban) is an ester of 3,4,4′-trichlorocarbanilide and belongs to the group of endocrine disruptors. It is widely used as a preservative and bactericide in a variety of personal care products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 101-20-2
Vapor Pressure <0.1 mmHg (25°C)
Flash Point >150°C
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula Cl2C6H3 NHCONHC6H4 Cl
MDL Number MFCD00013254
Melting Point 254°C to 256°C
Synonym 3,4,4′-Trichlorocarbanilide; 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea; TCC; Triclocarban
Type Certified Reference Material
Quantity 500 mg
Formula Weight 315.58
Grade Certified Reference Material
Show More Show Less