missing translation for 'onlineSavingsMsg'
Learn More

3,4,4'-Trichlorocarbanilide 98.0+%, TCI America™

Catalog No. T101525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T101525G 25 g
T1015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T101525G Supplier TCI America Supplier No. T101525G
Only null left

Chemical Identifiers

CAS 101-20-2
Molecular Formula C13H9Cl3N2O
Molecular Weight (g/mol) 315.58
MDL Number MFCD00013254
InChI Key ICUTUKXCWQYESQ-UHFFFAOYSA-N
PubChem CID 7547
ChEBI CHEBI:48347
IUPAC Name 3-(4-chlorophenyl)-1-(3,4-dichlorophenyl)urea
SMILES ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C(Cl)=C2)C=C1

Specifications

Color White
Quantity 25 g
UN Number 3077
Synonym 3,4,4′C-Trichlorodiphenylurea, Triclocarban, 1-(4-Chlorophenyl)-3-(3,4-dichlorophenyl)urea
Formula Weight 315.58
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,4′-Trichlorocarbanilide
EINECSNumber (9)-0920
RTECSNumber FE1250000
TSCA Yes