missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole 98.0+%, TCI America™
$158.44 - $799.68
Chemical Identifiers
| CAS | 1160294-96-1 |
|---|---|
| Molecular Formula | C36H26N2 |
| Molecular Weight (g/mol) | 486.618 |
| MDL Number | MFCD22571697 |
| InChI Key | WPQJQVZDUZXMTO-UHFFFAOYSA-N |
| PubChem CID | 59827995 |
| IUPAC Name | 4-phenyl-N-[4-(9-phenylcarbazol-3-yl)phenyl]aniline |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC5=C(C=C4)N(C6=CC=CC=C65)C7=CC=CC=C7 |
Chemical Identifiers
| 1160294-96-1 | |
| 486.618 | |
| WPQJQVZDUZXMTO-UHFFFAOYSA-N | |
| 4-phenyl-N-[4-(9-phenylcarbazol-3-yl)phenyl]aniline |
| C36H26N2 | |
| MFCD22571697 | |
| 59827995 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC5=C(C=C4)N(C6=CC=CC=C65)C7=CC=CC=C7 |
Specifications
| 1160294-96-1 | |
| White | |
| MFCD22571697 | |
| WPQJQVZDUZXMTO-UHFFFAOYSA-N | |
| 4-phenyl-N-[4-(9-phenylcarbazol-3-yl)phenyl]aniline | |
| 59827995 | |
| ≥98.0% (HPLC,N) | |
| 3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole |
| 180°C | |
| C36H26N2 | |
| 1 g | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC5=C(C=C4)N(C6=CC=CC=C65)C7=CC=CC=C7 | |
| 486.618 | |
| 486.62 | |
| Crystalline Powder |
Safety and Handling
TSCA : No