accessibility menu, dialog, popup

UserName

CAS RN 1160294-96-1

NHN

CAS RN 1160294-96-1

IUPAC Name: N-[4-(9-phenyl-9H-carbazol-3-yl)phenyl]-[1,1'-biphenyl]-4-amine
Synonyms: N-[4-(9-phenylcarbazol-3-yl)phenyl]-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 486.62
Molecular Formula: C36H26N2
InChi Key: WPQJQVZDUZXMTO-UHFFFAOYSA-N
SMILES: N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results