missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole 98.0+%, TCI America™

Catalog No. B48875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48875G 5 g
B48871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48875G Supplier TCI America Supplier No. B48875G

Chemical Identifiers

CAS 1160294-96-1
Molecular Formula C36H26N2
Molecular Weight (g/mol) 486.618
MDL Number MFCD22571697
InChI Key WPQJQVZDUZXMTO-UHFFFAOYSA-N
PubChem CID 59827995
IUPAC Name 4-phenyl-N-[4-(9-phenylcarbazol-3-yl)phenyl]aniline
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)NC3=CC=C(C=C3)C4=CC5=C(C=C4)N(C6=CC=CC=C65)C7=CC=CC=C7

Specifications

Melting Point 180°C
Color White
Quantity 5 g
Formula Weight 486.62
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-[4-(4-Biphenylylamino)phenyl]-9-phenylcarbazole