missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™
$121.16 - $410.10
Chemical Identifiers
| CAS | 214360-76-6 |
|---|---|
| Molecular Formula | C12H17BO3 |
| Molecular Weight (g/mol) | 220.075 |
| MDL Number | MFCD02093755 |
| InChI Key | MUKIFYQKIZOYKT-UHFFFAOYSA-N |
| Synonym | 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxyphenylboronic acid pinacol ester, 3-hydroxybenzeneboronic acid, pinacol ester, phenol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-hydroxyphenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, acmc-209fki, ksc497e2j |
| PubChem CID | 2734623 |
| IUPAC Name | 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O |
Chemical Identifiers
| 214360-76-6 | |
| 220.075 | |
| MUKIFYQKIZOYKT-UHFFFAOYSA-N | |
| 2734623 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O |
| C12H17BO3 | |
| MFCD02093755 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxyphenylboronic acid pinacol ester, 3-hydroxybenzeneboronic acid, pinacol ester, phenol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-hydroxyphenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, acmc-209fki, ksc497e2j | |
| 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Specifications
| 214360-76-6 | |
| C12H17BO3 | |
| 1 g | |
| MUKIFYQKIZOYKT-UHFFFAOYSA-N | |
| 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
| 2734623 | |
| ≥98.0% (GC) | |
| 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
| 99°C | |
| MFCD02093755 | |
| 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxyphenylboronic acid pinacol ester, 3-hydroxybenzeneboronic acid, pinacol ester, phenol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-hydroxyphenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, acmc-209fki, ksc497e2j | |
| B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O | |
| 220.075 | |
| 220.08 | |
| Crystalline Powder |
Safety and Handling
TSCA : No