missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 99°C |
| Quantity | 5 g |
| Formula Weight | 220.08 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
Safety and Handling
| TSCA | No |