missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol 98.0+%, TCI America™

Catalog No. T19531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19531G 1 g
T19535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19531G Supplier TCI America Supplier No. T19531G
Only null left

Chemical Identifiers

CAS 214360-76-6
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093755
InChI Key MUKIFYQKIZOYKT-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-hydroxyphenylboronic acid pinacol ester, 3-hydroxybenzeneboronic acid, pinacol ester, phenol, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-hydroxyphenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenol, 2-3-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, acmc-209fki, ksc497e2j
PubChem CID 2734623
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)O

Specifications

Melting Point 99°C
Quantity 1 g
Formula Weight 220.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol