missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™

Catalog No. p-7137586
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31171G 1 g
T31175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31171G Supplier TCI America Supplier No. T31171G

Chemical Identifiers

CAS 855738-89-5
Molecular Formula C18H20BNO2
Molecular Weight (g/mol) 293.173
MDL Number MFCD16996081
InChI Key ARVCVPGNHWNNAF-UHFFFAOYSA-N
Synonym 2-(9H-Carbazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (9H-Carbazol-3-yl)boronic Acid Pinacol Ester
PubChem CID 67070134
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)NC4=CC=CC=C43

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 1 g
Formula Weight 293.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole