missing translation for 'onlineSavingsMsg'
Learn More

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline 98.0+%, TCI America™

Catalog No. T30281G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30281G 1 g
T30285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30281G Supplier TCI America Supplier No. T30281G

Chemical Identifiers

CAS 210907-84-9
Molecular Formula C12H18BNO2
Molecular Weight (g/mol) 219.09
MDL Number MFCD03453668
InChI Key YMXIIVIQLHYKOT-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-aminophenylboronic acid pinacol ester, benzenamine, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-aminophenylboronic acid, pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-aminobenzeneboronic acid pinacol ester, 3-aminophenylboronic acid, pinacol cyclic ester, 3-aminophenylboronicacidpinacolester, apbe, amtb060
PubChem CID 2734655
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(N)=C1

Specifications

Melting Point 94°C
Quantity 1 g
Formula Weight 219.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline