accessibility menu, dialog, popup

UserName

CAS RN 210907-84-9

IUPAC Name: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 219.09
Molecular Formula: C12H18BNO2
InChi Key: YMXIIVIQLHYKOT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC(N)=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)