missing translation for 'onlineSavingsMsg'
Learn More

3'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide 98.0+%, TCI America™

Catalog No. T32161G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T32161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T32161G Supplier TCI America Supplier No. T32161G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 480424-93-9
Molecular Formula C14H20BNO3
Molecular Weight (g/mol) 261.128
MDL Number MFCD03789261
InChI Key CZFSGYCLOCCASM-UHFFFAOYSA-N
Synonym 3-Acetamidophenylboronic Acid Pinacol Ester, 2-(3-Acetamidophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 2773989
IUPAC Name N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NC(=O)C

Specifications

Melting Point 190°C
Color White-Yellow
Quantity 1 g
Formula Weight 261.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acetanilide