missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Trifluoromethylphenyl)propionic Acid 98.0+%, TCI America™

Catalog No. T29311G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29311G 1 g
T29315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29311G Supplier TCI America Supplier No. T29311G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 585-50-2
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.18
MDL Number MFCD00090487
InChI Key YLTJJMIWCCJIHI-UHFFFAOYSA-N
Synonym 3-3-trifluoromethyl phenyl propanoic acid, 3-3-trifluoromethylphenyl propionic acid, benzenepropanoic acid, 3-trifluoromethyl, 3-trifluoromethyl hydrocinnamic acid, unii-4u44xhh79h, 3-3'-trifluoromethylphenyl-propionic acid, 3-3-trifluoromethyl phenyl propionic acid, cinacalcet m5, m-trifluoromethyl hydrocinnamic acid
PubChem CID 2777683
IUPAC Name 3-[3-(trifluoromethyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=CC(=CC=C1)C(F)(F)F

Specifications

Melting Point 35°C
Color White-Yellow
Boiling Point 149°C
Quantity 1 g
Formula Weight 218.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-(3-Trifluoromethylphenyl)propionic Acid