accessibility menu, dialog, popup

UserName

CAS RN 585-50-2

IUPAC Name: 3-[3-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[3-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: YLTJJMIWCCJIHI-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(=CC=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results