missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide] 98.0+%, TCI America™

Catalog No. m27911g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27911G 1 g
M27915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27911G Supplier TCI America Supplier No. M27911G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 889617-36-1
Molecular Formula C31H42Br2N4
Molecular Weight (g/mol) 630.513
InChI Key ZZGWEIDPDBDPOH-UHFFFAOYSA-L
PubChem CID 78198750
IUPAC Name 1-[2,6-di(propan-2-yl)phenyl]-3-[[3-[2,6-di(propan-2-yl)phenyl]imidazol-1-ium-1-yl]methyl]imidazol-3-ium;dibromide
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N2C=C[N+](=C2)C[N+]3=CN(C=C3)C4=C(C=CC=C4C(C)C)C(C)C.[Br-].[Br-]

Specifications

Melting Point 277°C
Color White
Quantity 1 g
Formula Weight 630.51
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Methylenebis[1-(2,6-diisopropylphenyl)-3-imidazolium Bromide]