accessibility menu, dialog, popup

UserName

CH3CH3H3CCH3NNNNCH3H3CCH3CH3BrBr

CAS RN 889617-36-1

IUPAC Name: 1-[2,6-bis(propan-2-yl)phenyl]-3-({1-[2,6-bis(propan-2-yl)phenyl]-1H-imidazol-3-ium-3-yl}methyl)-1H-imidazol-3-ium dibromide
Synonyms: 3-(2,6-diisopropylphenyl)-1-{[3-(2,6-diisopropylphenyl)imidazol-1-ium-1-yl]methyl}imidazol-1-ium dibromide
Molecular Weight (g/mol): 630.51
Molecular Formula: C31H42Br2N4
InChi Key: ZZGWEIDPDBDPOH-UHFFFAOYSA-L
SMILES: [Br-].[Br-].CC(C)C1=CC=CC(C(C)C)=C1N1C=C[N+](C[N+]2=CN(C=C2)C2=C(C=CC=C2C(C)C)C(C)C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results