missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)-1H-pyrazole, 97%

Catalog No. p-7033339
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5043503 1 g
AAH5043506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5043503 Supplier Thermo Scientific Chemicals Supplier No. H5043503
Only null left

CAS: 144026-74-4 | C10H10N2O | 174.203 g/mol

3-(3-Methoxyphenyl)-1H-pyrazole is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(3-Methoxyphenyl)-1H-pyrazole is used as pharmaceutical intermediates.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.

Chemical Identifiers

CAS 144026-74-4
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.203
MDL Number MFCD01940432
InChI Key JAZQIOZMPNVSII-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl-1h-pyrazole, 5-3-methoxyphenyl-1h-pyrazole, 1h-pyrazole, 3-3-methoxyphenyl, 3-3-methoxyphenyl pyrazole, 3-3-methoxy-phenyl-1h-pyrazole, 3-3-methoxyphenyl-2h-pyrazole, acmc-20ankv, 1-methoxy-3-pyrazol-3-ylbenzene, 3-methoxy-1-pyrazol-3-ylbenzene, 1h-pyrazole,3-3-methoxyphenyl
PubChem CID 2736766
IUPAC Name 5-(3-methoxyphenyl)-1H-pyrazole
SMILES COC1=CC=CC(=C1)C2=CC=NN2

Specifications

Melting Point 90°C to 91°C
Quantity 1 g
Formula Weight 174.2
Percent Purity 97%
Chemical Name or Material 3-(3-Methoxyphenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only