missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Iminodipropionic Acid 98.0+%, TCI America™

$465.89 - $1609.44

Chemical Identifiers

CAS 505-47-5
Molecular Formula C6H11NO4
Molecular Weight (g/mol) 161.16
MDL Number MFCD01311684
InChI Key TXPKUUXHNFRBPS-UHFFFAOYSA-N
PubChem CID 10456
IUPAC Name 3-[(2-carboxyethyl)amino]propanoic acid
SMILES OC(=O)CCNCCC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I08231G
View Documents
TCI America
I08231G
1 g
Each for $465.89
Only null left
 
I08235G
View Documents
TCI America
I08235G
5 g
Each for $1,609.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

505-47-5
161.16
TXPKUUXHNFRBPS-UHFFFAOYSA-N
3-[(2-carboxyethyl)amino]propanoic acid
C6H11NO4
MFCD01311684
10456
OC(=O)CCNCCC(O)=O

Specifications

156°C
White
MFCD01311684
OC(=O)CCNCCC(O)=O
1 g
10456
≥98.0% (GC,T)
3,3′-Iminodipropionic Acid
505-47-5
C6H11NO4
TXPKUUXHNFRBPS-UHFFFAOYSA-N
3-[(2-carboxyethyl)amino]propanoic acid
161.16
161.16
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents