CAS RN 505-47-5
CAS RN 505-47-5
IUPAC Name:
3-[(2-carboxyethyl)amino]propanoic acid
Synonyms:
3-[(2-carboxyethyl)amino]propanoic acid
Molecular Weight (g/mol):
161.16
Molecular Formula:
C6H11NO4
InChi Key:
TXPKUUXHNFRBPS-UHFFFAOYSA-N
SMILES:
OC(=O)CCNCCC(O)=O
1
–
1
of
1
results
1
3,3'-Iminodipropionic Acid 98.0+%, TCI America™
CAS: 505-47-5 Molecular Formula: C6H11NO4 Molecular Weight (g/mol): 161.16 MDL Number: MFCD01311684 InChI Key: TXPKUUXHNFRBPS-UHFFFAOYSA-N PubChem CID: 10456 IUPAC Name: 3-[(2-carboxyethyl)amino]propanoic acid SMILES: OC(=O)CCNCCC(O)=O