missing translation for 'onlineSavingsMsg'
Learn More

3,3'-(Ethylenedithio)dipropionitrile 97.0+%, TCI America™

Catalog No. E06731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E06731G 1 g
E06735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E06731G Supplier TCI America Supplier No. E06731G
Only null left

Chemical Identifiers

CAS 86180-54-3
Molecular Formula C8H12N2S2
Molecular Weight (g/mol) 200.32
MDL Number MFCD00475273
InChI Key SJYFOHJCZZEPNC-UHFFFAOYSA-N
Synonym 1,2-Bis(2-cyanoethylthio)ethane, 4,7-Dithiadecanedinitrile
PubChem CID 99335
IUPAC Name 3-({2-[(2-cyanoethyl)sulfanyl]ethyl}sulfanyl)propanenitrile
SMILES N#CCCSCCSCCC#N

Specifications

Melting Point 93°C
Color White
Quantity 1 g
Formula Weight 200.32
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-(Ethylenedithio)dipropionitrile
RTECSNumber TZ4685000
TSCA No