missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dithiodipropionic Acid 99.0+%, TCI America™
$44.28 - $261.82
Chemical Identifiers
| CAS | 1119-62-6 |
|---|---|
| Molecular Formula | C6H10O4S2 |
| Molecular Weight (g/mol) | 210.262 |
| MDL Number | MFCD00002780 |
| InChI Key | YCLSOMLVSHPPFV-UHFFFAOYSA-N |
| Synonym | 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid |
| PubChem CID | 95116 |
| IUPAC Name | 3-(2-carboxyethyldisulfanyl)propanoic acid |
| SMILES | C(CSSCCC(=O)O)C(=O)O |
Chemical Identifiers
| 1119-62-6 | |
| 210.262 | |
| YCLSOMLVSHPPFV-UHFFFAOYSA-N | |
| 95116 | |
| C(CSSCCC(=O)O)C(=O)O |
| C6H10O4S2 | |
| MFCD00002780 | |
| 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid | |
| 3-(2-carboxyethyldisulfanyl)propanoic acid |
Specifications
| 1119-62-6 | |
| White | |
| MFCD00002780 | |
| 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid | |
| C(CSSCCC(=O)O)C(=O)O | |
| 210.262 | |
| 210.26 | |
| Crystalline Powder |
| 157°C | |
| C6H10O4S2 | |
| 25 g | |
| YCLSOMLVSHPPFV-UHFFFAOYSA-N | |
| 3-(2-carboxyethyldisulfanyl)propanoic acid | |
| 95116 | |
| ≥99.0% (GC,T) | |
| 3,3′-Dithiodipropionic Acid |
Safety and Handling
TSCA : Yes