missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiodipropionic Acid 99.0+%, TCI America™

Catalog No. D094725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D094725G 25 g
D0947100G 100 g
D0947500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D094725G Supplier TCI America Supplier No. D094725G

Chemical Identifiers

CAS 1119-62-6
Molecular Formula C6H10O4S2
Molecular Weight (g/mol) 210.262
MDL Number MFCD00002780
InChI Key YCLSOMLVSHPPFV-UHFFFAOYSA-N
Synonym 3,3'-dithiodipropionic acid, dithiodipropionic acid, propanoic acid, 3,3'-dithiobis, 3,3-dithiodipropionic acid, 3,3'-dithiobis dipropionic acid, unii-42op5b2ni6, 3,3'-dithiobispropionic acid, propionic acid, 3,3'-dithiodi, 3,3'-disulfanediyldipropanoic acid, 3-2-carboxyethyl disulfanyl propanoic acid
PubChem CID 95116
IUPAC Name 3-(2-carboxyethyldisulfanyl)propanoic acid
SMILES C(CSSCCC(=O)O)C(=O)O

Specifications

Melting Point 157°C
Color White
Quantity 25 g
Formula Weight 210.26
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dithiodipropionic Acid