missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenyl-L-alanine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5216906

 View more versions of this product

Catalog No. AAH5216906


Only null left

Description

Description

3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3,3-Diphenyl-L-alanine
C15H15NO2
UN2811
PECGVEGMRUZOML-AWEZNQCLSA-N
(2S)-2-amino-3,3-diphenylpropanoic acid
241.29
241.29
149597-92-2
MFCD01631989
3,3-diphenyl-l-alanine, s-2-amino-3,3-diphenylpropanoic acid, l-3,3-diphenylalanine, 3,3-diphenylalanine, 2s-2-amino-3,3-diphenylpropanoic acid, s-2-amino-3,3-diphenyl-propionic acid, 2s-2-amino-3,3-diphenyl-propanoic acid, d-3,3'-biphenylalanine, beta-phenylphenylalanine, pubchem18701
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)N
5 g
162977
95%

Chemical Identifiers

149597-92-2
241.29
PECGVEGMRUZOML-AWEZNQCLSA-N
162977
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)N
C15H15NO2
MFCD01631989
3,3-diphenyl-l-alanine, s-2-amino-3,3-diphenylpropanoic acid, l-3,3-diphenylalanine, 3,3-diphenylalanine, 2s-2-amino-3,3-diphenylpropanoic acid, s-2-amino-3,3-diphenyl-propionic acid, 2s-2-amino-3,3-diphenyl-propanoic acid, d-3,3'-biphenylalanine, beta-phenylphenylalanine, pubchem18701
(2S)-2-amino-3,3-diphenylpropanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P310-P330-P501c

H301

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only