missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenyl-L-alanine, 95%, Thermo Scientific Chemicals

Catalog No. p-7034483
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5216903 1 g
AAH5216906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5216903 Supplier Thermo Scientific Chemicals Supplier No. H5216903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 149597-92-2 | Molecular Formula: C15H15NO2 | Molecular Weight: 241.29 g/mol

3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3-Diphenyl-L-alanine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 149597-92-2
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD01631989
InChI Key PECGVEGMRUZOML-AWEZNQCLSA-N
Synonym 3,3-diphenyl-l-alanine, s-2-amino-3,3-diphenylpropanoic acid, l-3,3-diphenylalanine, 3,3-diphenylalanine, 2s-2-amino-3,3-diphenylpropanoic acid, s-2-amino-3,3-diphenyl-propionic acid, 2s-2-amino-3,3-diphenyl-propanoic acid, d-3,3'-biphenylalanine, beta-phenylphenylalanine, pubchem18701
PubChem CID 162977
IUPAC Name (2S)-2-amino-3,3-diphenylpropanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)C(C(=O)O)N

Specifications

UN Number UN2811
Quantity 1 g
Formula Weight 241.29
Percent Purity 95%
Chemical Name or Material 3,3-Diphenyl-L-alanine
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only