missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,3'-Dimethyltriphenylamine 98.0+%, TCI America™
$158.44 - $480.92
Chemical Identifiers
| CAS | 13511-11-0 |
|---|---|
| Molecular Formula | C20H19N |
| Molecular Weight (g/mol) | 273.379 |
| MDL Number | MFCD23115649 |
| InChI Key | ZCAWQFNYHFHEPZ-UHFFFAOYSA-N |
| Synonym | Bis(3-methylphenyl)phenylamine, N,N-Di-m-tolylaniline, Phenyl(di-m-tolyl)amine |
| PubChem CID | 18326012 |
| IUPAC Name | 3-methyl-N-(3-methylphenyl)-N-phenylaniline |
| SMILES | CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=CC(=C3)C |
Chemical Identifiers
| 13511-11-0 | |
| 273.379 | |
| ZCAWQFNYHFHEPZ-UHFFFAOYSA-N | |
| 18326012 | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=CC(=C3)C |
| C20H19N | |
| MFCD23115649 | |
| Bis(3-methylphenyl)phenylamine, N,N-Di-m-tolylaniline, Phenyl(di-m-tolyl)amine | |
| 3-methyl-N-(3-methylphenyl)-N-phenylaniline |
Specifications
| 13511-11-0 | |
| White-Yellow | |
| MFCD23115649 | |
| Bis(3-methylphenyl)phenylamine, N,N-Di-m-tolylaniline, Phenyl(di-m-tolyl)amine | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=CC=CC(=C3)C | |
| 273.379 | |
| 273.38 | |
| Crystalline Powder |
| 72°C | |
| C20H19N | |
| 1 g | |
| ZCAWQFNYHFHEPZ-UHFFFAOYSA-N | |
| 3-methyl-N-(3-methylphenyl)-N-phenylaniline | |
| 18326012 | |
| ≥98.0% (GC) | |
| 3,3′-Dimethyltriphenylamine |
Safety and Handling
TSCA : No