accessibility menu, dialog, popup

UserName

CH3NCH3

CAS RN 13511-11-0

IUPAC Name: 3-methyl-N-(3-methylphenyl)-N-phenylaniline
Synonyms: 3-methyl-N-(3-methylphenyl)-N-phenylaniline
Molecular Weight (g/mol): 273.38
Molecular Formula: C20H19N
InChi Key: ZCAWQFNYHFHEPZ-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N(C1=CC=CC=C1)C1=CC=CC(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results