missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyloctane 98.0+%, TCI America™

Catalog No. D12131ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12131ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D12131ML Supplier TCI America Supplier No. D12131ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4110-44-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00048874
InChI Key DBULLUBYDONGLT-UHFFFAOYSA-N
Synonym octane, 3,3-dimethyl, 3,3-dimethyl octane, acmc-209jii
PubChem CID 138117
IUPAC Name 3,3-dimethyloctane
SMILES CCCCCC(C)(C)CC

Specifications

Color Colorless
Boiling Point 161°C
Quantity 1 mL
UN Number 3295
Formula Weight 142.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyloctane
EINECSNumber (2)-0010
TSCA No