accessibility menu, dialog, popup

UserName

CH3CH3H3CH3C

CAS RN 4110-44-5

IUPAC Name: 3,3-dimethyloctane
Synonyms: octane, 3,3-dimethyl-
Molecular Weight (g/mol): 142.29
Molecular Formula: C10H22
InChi Key: DBULLUBYDONGLT-UHFFFAOYSA-N
SMILES: CCCCCC(C)(C)CC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)