missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyloctane, 98%

Catalog No. AAB2152003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2152003 1 g
AAB2152006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2152003 Supplier Thermo Scientific Chemicals Supplier No. B2152003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4110-44-5 | C10H22 | 142.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4110-44-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00048874
InChI Key DBULLUBYDONGLT-UHFFFAOYSA-N
Synonym octane, 3,3-dimethyl, 3,3-dimethyl octane, acmc-209jii
PubChem CID 138117
IUPAC Name 3,3-dimethyloctane
SMILES CCCCCC(C)(C)CC

Specifications

Density 0.74
Boiling Point 161°C
Quantity 1 g
UN Number UN3295
Formula Weight 142.29
Percent Purity 98%
Chemical Name or Material 3,3-Dimethyloctane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only