missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethyldiphenylamine, 98%

Catalog No. p-7025999
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2176503 1 g
AAB2176506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2176503 Supplier Thermo Scientific Chemicals Supplier No. B2176503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-13-1 | C14H15N | 197.281 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626-13-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00059315
InChI Key CWVPIIWMONJVGG-UHFFFAOYSA-N
Synonym di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
PubChem CID 7016139
IUPAC Name 3-methyl-N-(3-methylphenyl)aniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC(=C2)C

Specifications

Density 1.04
Boiling Point 200°C (24 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6225
Quantity 1 g
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material 3,3'-Dimethyldiphenylamine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only