missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethylazobenzene 98.0+%, TCI America™

Catalog No. D06765G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06765G 5 g
D06761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D06765G Supplier TCI America Supplier No. D06765G
Only null left

Chemical Identifiers

CAS 588-04-5
Molecular Formula C14H14N2
Molecular Weight (g/mol) 210.28
MDL Number MFCD00048080
InChI Key HPSZIXYLILUGIP-UHFFFAOYSA-N
Synonym m-Azotoluene
PubChem CID 11491
IUPAC Name bis(3-methylphenyl)diazene
SMILES CC1=CC(=CC=C1)N=NC2=CC=CC(=C2)C

Specifications

Melting Point 53°C
Quantity 5 g
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Dimethylazobenzene