accessibility menu, dialog, popup

UserName

CAS RN 588-04-5

CH3NNH3C(E/Z)

CAS RN 588-04-5

IUPAC Name: bis(3-methylphenyl)diazene
Synonyms: bis(3-methylphenyl)diazene
Molecular Weight (g/mol): 210.28
Molecular Formula: C14H14N2
InChi Key: HPSZIXYLILUGIP-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1)N=NC1=CC(C)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results