missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-1-butyne 96.0+%, TCI America™

Catalog No. p-7118858
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B111410ML 10 mL
B1114100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B111410ML Supplier TCI America Supplier No. B111410ML
Only null left

Chemical Identifiers

CAS 917-92-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008852
InChI Key PPWNCLVNXGCGAF-UHFFFAOYSA-N
PubChem CID 13512
IUPAC Name 3,3-dimethylbut-1-yne
SMILES CC(C)(C)C#C

Specifications

Color Colorless
Boiling Point 38°C
Quantity 10 mL
UN Number 3295
Synonym 3,3-dimethyl-1-butyne, tert-butylacetylene, t-butylacetylene, 3,3-dimethylbutyne, 1-butyne, 3,3-dimethyl, t-butyl acetylene, 3,3,3-trimethylpropyne, 1-butyne,3,3-dimethyl, tertbutylacetylene, tert-butylacetyiene
Formula Weight 82.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,3-Dimethyl-1-butyne
TSCA No
Recommended Storage Refrigerator