missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Diethylbenzidine Dihydrochloride 97.0+%, TCI America™

Catalog No. D04811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D04811G Supplier TCI America Supplier No. D04811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76787-89-8
Molecular Formula C16H22Cl2N2
Molecular Weight (g/mol) 313.266
MDL Number MFCD00060214
InChI Key DQPPOEIMMLEMPW-UHFFFAOYSA-N
Synonym 4,4′C-Diamino-3,3′C-diethylbiphenyl Dihydrochloride
PubChem CID 18968080
IUPAC Name 4-(4-amino-3-ethylphenyl)-2-ethylaniline;dihydrochloride
SMILES CCC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)CC)N.Cl.Cl

Specifications

Quantity 1 g
Formula Weight 313.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′-Diethylbenzidine Dihydrochloride